Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178005.4 | NP_821072.1 | 2 | 755 | ||
GenBank mRNA | AK036155.1 | - | 2 | 750 | ||
AK043173.1 | - | 2 | 611 | |||
AK043784.1 | - | 2 | 689 | |||
AK044705.1 | - | 2 | 757 | |||
AK046774.1 | - | 2 | 756 | |||
AK046818.1 | - | 2 | 650 | |||
AK049920.1 | - | 2 | 650 | |||
AK122278.1 | - | 2 | 741 | |||
AY182027.1 | - | 1 | 327 |