Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178358.3 | NP_848135.1 | 3 | 770 | ||
GenBank mRNA | AK142526.1 | - | 3 | 827 | ||
AY253666.1 | - | 3 | 826 | |||
BC051434.1 | - | 3 | 770 | 6389932 |