Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178387.3 | NP_848474.2 | 13 | 1934 | ||
GenBank mRNA | AK006916.1 | - | 12 | 1613 |