Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286205.1 | NP_001273134.1 | 3 | 394 | ||
NM_001286208.1 | NP_001273137.1 | 3 | 394 | |||
NM_001286209.1 | NP_001273138.1 | 3 | 286 | |||
NM_001481.2 | NP_001472.1 | 3 | 271 | |||
XM_005256304.4 | XP_005256361.1 | 4 | 926 | |||
XM_005256309.4 | XP_005256366.1 | 5 | 651 | |||
XM_006721175.3 | XP_006721238.1 | 3 | 324 | |||
XM_011522990.2 | XP_011521292.1 | 3 | 345 | |||
XM_011522991.1 | XP_011521293.1 | 4 | 500 | |||
XM_011522992.2 | XP_011521294.1 | 2 | 293 | |||
XM_017023122.1 | XP_016878611.1 | 5 | 651 | |||
XM_017023123.1 | XP_016878612.1 | 3 | 350 | |||
XM_017023124.1 | XP_016878613.1 | 3 | 192 | |||
XM_017023125.1 | XP_016878614.1 | 3 | 324 | |||
GenBank mRNA | AF050079.1 | - | 3 | 270 | ||
AK293958.1 | - | 3 | 301 | |||
AK294050.1 | - | 2 | 120 | |||
AK297857.1 | - | 3 | 254 | |||
AK303505.1 | - | 3 | 327 | |||
AK304731.1 | - | 3 | 188 | |||
AK315261.1 | - | 3 | 219 | |||
BC104785.1 | - | 3 | 256 | 8143788 | ||
BC112121.1 | - | 3 | 256 | 8327589 | ||
BX647947.1 | - | 3 | 312 | |||
CR749499.1 | - | 2 | 304 | |||
DC370728.1 | - | 3 | 272 | |||
DC371501.1 | - | 3 | 329 |