Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bend7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190400.1 | NP_001177329.1 | 2 | 484 | ||
NM_178663.4 | NP_848778.1 | 2 | 484 | |||
GenBank mRNA | AK030428.1 | - | 2 | 171 | ||
AK042822.1 | - | 2 | 482 | |||
AK053896.1 | - | 2 | 486 | |||
BC096430.1 | - | 2 | 156 | 5041388 | ||
BC119370.1 | - | 2 | 123 | 8734119 | ||
BC144864.1 | - | 2 | 123 | 9053412 |