Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164562.1 | NP_001158034.1 | 2 | 225 | ||
NM_178677.4 | NP_848792.2 | 3 | 350 | |||
GenBank mRNA | AK029985.1 | - | 3 | 281 | ||
AK031125.1 | - | 2 | 222 | |||
AK044249.1 | - | 2 | 225 | |||
BB661643.1 | - | 3 | 350 | |||
BC046289.1 | - | 2 | 180 | 6469221 |