Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184706.1 | NP_001171635.1 | 13 | 1983 | ||
NM_001184708.1 | NP_001171637.1 | 10 | 1709 | |||
NM_001184709.1 | NP_001171638.1 | 11 | 1607 | |||
NM_001184710.1 | NP_001171639.1 | 10 | 1483 | |||
NM_001184711.1 | NP_001171640.1 | 11 | 1590 | |||
NM_178667.4 | NP_848782.2 | 14 | 1984 | |||
GenBank mRNA | AK029055.1 | - | 13 | 1956 | ||
AK033356.1 | - | 11 | 1571 | |||
AK044784.1 | - | 10 | 1711 | |||
AK053881.1 | - | 13 | 1942 | |||
AK079624.1 | - | 14 | 1912 | |||
AK158829.1 | - | 10 | 1479 | |||
BC092230.1 | - | 11 | 1553 |