Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000016.5 | NP_000007.1 | 8 | 1075 | ||
NM_001127328.2 | NP_001120800.1 | 8 | 1087 | |||
NM_001286042.1 | NP_001272971.1 | 7 | 987 | |||
NM_001286043.1 | NP_001272972.1 | 9 | 1174 | |||
NM_001286044.1 | NP_001272973.1 | 5 | 805 | |||
GenBank mRNA | AF251043.1 | - | 8 | 654 | ||
AK296045.1 | - | 5 | 385 | |||
AK301005.1 | - | 5 | 543 | |||
AK301039.1 | - | 8 | 731 | |||
AK301717.1 | - | 7 | 579 | |||
AK311022.1 | - | 7 | 557 | |||
AK312629.1 | - | 8 | 665 | |||
AK315946.1 | - | 7 | 628 | |||
BC005377.1 | - | 8 | 669 | 3934299 | ||
BX647734.1 | - | 9 | 813 | |||
KU177876.1 | - | 8 | 634 | |||
KU177877.1 | - | 7 | 546 | |||
KU177878.1 | - | 4 | 277 | |||
M16827.1 | - | 8 | 652 |