Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001609.3 | NP_001600.1 | 5 | 772 | ||
XM_017016194.1 | XP_016871683.1 | 4 | 613 | |||
GenBank mRNA | AK298638.1 | - | 4 | 601 | ||
AK314241.1 | - | 5 | 668 | |||
AL831821.2 | - | 5 | 641 | |||
BC013756.1 | - | 5 | 667 | 4706281 | ||
KU177879.1 | - | 5 | 655 | |||
KU177880.1 | - | 6 | 963 | |||
U12778.1 | - | 5 | 670 |