Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098.2 | NP_001089.1 | 8 | 1020 | ||
XM_017028812.1 | XP_016884301.1 | 8 | 1334 | |||
GenBank mRNA | AK293562.1 | - | 4 | 242 | ||
AK296260.1 | - | 9 | 985 | |||
AK297213.1 | - | 8 | 895 | |||
AK302691.1 | - | 9 | 970 | |||
AK314845.1 | - | 8 | 1028 | |||
BC014092.2 | - | 8 | 1020 | 4328775 | ||
BC026196.2 | - | 8 | 1023 | 5264179 | ||
CR456365.1 | - | 8 | 1012 | |||
CR536568.1 | - | 8 | 998 | |||
GQ472196.1 | - | 8 | 1023 | |||
U80040.1 | - | 8 | 1018 |