Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302489.1 | NP_001289418.1 | 6, 7 | 663 | ||
NM_001302490.1 | NP_001289419.1 | 5, 6 | 595 | |||
NM_001302492.1 | NP_001289421.1 | 5, 6 | 620 | |||
NM_001610.3 | NP_001601.1 | 6, 7 | 691 | |||
GenBank mRNA | AK298439.1 | - | 6, 7 | 666 | ||
AK299382.1 | - | 6, 7 | 620 | |||
AK300569.1 | - | 5, 6 | 619 | |||
AK300911.1 | - | 5, 6 | 592 | |||
AK301860.1 | - | 5, 6 | 553 | |||
AK303894.1 | - | 5, 6 | 587 | |||
AK308512.1 | - | 4, 5 | 910 | |||
AK311431.1 | - | 5, 6 | 585 | |||
AY007137.1 | - | 3, 4 | 223 | |||
BC003160.1 | - | 6, 7 | 636 | 2957907 | ||
BC093010.1 | - | 6, 7 | 649 | 30332133 | ||
X12548.1 | - | 6, 7 | 645 |