Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109.4 | NP_001100.3 | 2 | 170 | ||
NM_001164489.1 | NP_001157961.1 | 2 | 170 | |||
XM_011539117.2 | XP_011537419.2 | 2 | 156 | |||
XM_017015465.1 | XP_016870954.1 | 2 | 156 | |||
XM_017015466.1 | XP_016870955.1 | 2 | 158 | |||
GenBank mRNA | AK304324.1 | - | 2 | 327 | ||
AK312834.1 | - | 2 | 135 | |||
BC064500.1 | - | 2 | 117 | |||
BC115404.1 | - | 2 | 89 | 40075505 | ||
BM146234.1 | - | 2 | 136 | |||
D26579.1 | - | 2 | 93 | |||
DB142906.1 | - | 1 | 49 | |||
DC401057.1 | - | 2 | 171 | |||
DC408787.1 | - | 2 | 163 |