Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005161.4 | NP_005152.1 | 1 | 499 | ||
NR_027991.1 | 1 | 499 | ||||
GenBank mRNA | AK074514.1 | - | 1 | 298 | ||
AK075252.1 | - | 1 | 247 | |||
AK097232.1 | - | 1 | 24 | |||
AK315542.1 | - | 1 | 296 | |||
AL548962.3 | - | 1 | 231 | |||
BC032688.1 | - | 1 | 281 | 5241995 | ||
X89271.1 | - | 1 | 359 |
Set Membership: |
![]() |