Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000030.2 | NP_000021.1 | 10 | 1160 | ||
GenBank mRNA | AB292648.1 | - | 10 | 1061 | ||
AF191687.1 | - | 2 | 817 | |||
AF348451.1 | - | 10 | 951 | |||
AK292754.1 | - | 10 | 1081 | |||
AK315331.1 | - | 10 | 1108 | |||
BC132819.1 | - | 10 | 1157 | 40146841 | ||
D13368.1 | - | 10 | 1147 | |||
X53414.1 | - | 10 | 1161 | |||
X56092.1 | - | 10 | 1060 |