Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000697.2 | NP_000688.2 | 13 | 1822 | ||
XM_011523780.2 | XP_011522082.1 | 12 | 3889 | |||
GenBank mRNA | AF143883.1 | - | 2 | 487 | ||
BC069557.1 | - | 13 | 1800 | 7262463 | ||
M35418.1 | - | 13 | 1810 | |||
M58704.1 | - | 13 | 1802 | |||
M62982.1 | - | 13 | 1822 |