Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287214.1 | NP_001274143.1 | 1 | 363 | ||
NM_001287216.1 | NP_001274145.1 | 1 | 363 | |||
NM_001634.5 | NP_001625.2 | 1 | 363 | |||
NR_109768.1 | 1 | 363 | ||||
GenBank mRNA | AJ420527.1 | - | 1 | 338 | ||
AK300330.1 | - | 1 | 362 | |||
AK302761.1 | - | 1 | 364 | |||
AL832698.1 | - | 1 | 362 | |||
AU134914.1 | - | 1 | 363 | |||
BC000171.2 | - | 1 | 136 | 2900587 | ||
BX640599.1 | - | 1 | 363 | |||
M21154.1 | - | 1 | 289 |