Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BIN1 SIRNA Assays ›
Gene Symbol:
BIN1
Gene Name:
bridging integrator 1
Gene Aliases:
AMPH2, AMPHL, SH3P9
Chromosome Location:
Chr.2: 127048023 - 127107400 on Build GRCh38
UniGene ID:
Hs.193163
Species:
Human
siRNA ID
HSS100470
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320632.1
NP_001307561.1
7
943
NM_001320633.1
NP_001307562.1
8
1036
NM_001320634.1
NP_001307563.1
7
709
NM_001320640.1
NP_001307569.1
8
1036
NM_001320641.1
NP_001307570.1
7
943
NM_001320642.1
NP_001307571.1
8
1530
NM_004305.3
NP_004296.1
7
943
NM_139343.2
NP_647593.1
8
1036
NM_139344.2
NP_647594.1
8
1036
NM_139345.2
NP_647595.1
8
1036
NM_139346.2
NP_647596.1
7
943
NM_139347.2
NP_647597.1
7
943
NM_139348.2
NP_647598.1
7
943
NM_139349.2
NP_647599.1
7
943
NM_139350.2
NP_647600.1
7
943
NM_139351.2
NP_647601.1
7
943
XM_005263642.3
XP_005263699.1
8
1109
XM_005263643.3
XP_005263700.1
7
1016
XM_005263645.2
XP_005263702.1
7
500
XM_006712425.3
XP_006712488.1
8
1109
XM_006712426.3
XP_006712489.1
8
1109
XM_006712427.2
XP_006712490.1
7
500
XM_006712428.3
XP_006712491.1
8
1109
XM_006712429.3
XP_006712492.1
8
1109
XM_006712431.3
XP_006712494.1
8
1109
XM_006712432.3
XP_006712495.1
8
1109
XM_006712433.3
XP_006712496.1
8
1109
XM_006712434.3
XP_006712497.1
8
1109
XM_011510975.2
XP_011509277.1
8
1742
XM_017003819.1
XP_016859308.1
8
923
XM_017003820.1
XP_016859309.1
7
1016
XM_017003821.1
XP_016859310.1
8
1109
XM_017003822.1
XP_016859311.1
7
1016
XM_017003823.1
XP_016859312.1
7
500
XM_017003824.1
XP_016859313.1
7
500
XM_017003825.1
XP_016859314.1
8
923
XM_017003826.1
XP_016859315.1
7
500
XM_017003827.1
XP_016859316.1
8
1109
XM_017003828.1
XP_016859317.1
8
1109
GenBank mRNA
AF001383.1
-
7
730
AF004015.1
-
8
652
AF043898.1
-
7
559
AF043899.1
-
8
652
AF043900.1
-
7
559
AF043901.1
-
8
652
AF068914.1
-
7
619
AF068915.1
-
7
619
AF068916.1
-
7
619
AF068917.1
-
7
619
AF068918.1
-
7
619
AF070576.1
-
4
258
AK295300.1
-
8
754
AK301153.1
-
7
709
AL537027.3
-
8
790
AL713697.1
-
8
997
AW411140.1
-
3
214
BC004101.2
-
7
619
3688734
BT006865.1
-
7
559
U68485.1
-
7
605
U87558.1
-
8
673
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
nucleus organization
cell proliferation
viral process
regulation of endocytosis
muscle cell differentiation
positive regulation of apoptotic process
regulation of neuron differentiation
positive regulation of astrocyte differentiation
lipid tube assembly
regulation of cell cycle arrest
protein binding
identical protein binding
tau protein binding
actin filament binding
RNA polymerase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0