Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001118887.1 | NP_001112359.1 | 6, 7 | 1368 | ||
NM_001118888.1 | NP_001112360.1 | 5, 6 | 1215 | |||
NM_001147.2 | NP_001138.1 | 6, 7 | 1371 | |||
XM_017013318.1 | XP_016868807.1 | 5, 6 | 1369 | |||
GenBank mRNA | AB009865.1 | - | 6, 7 | 1067 | ||
AF004327.1 | - | 6, 7 | 1391 | |||
AF187858.1 | - | 5, 6 | 906 | |||
AF218015.1 | - | 6, 7 | 1122 | |||
AK075219.1 | - | 6, 7 | 1349 | |||
AK225698.1 | - | 6, 7 | 1350 | |||
AK290070.1 | - | 6, 7 | 1353 | |||
AK310171.1 | - | 6, 7 | 1350 | |||
AK312541.1 | - | 6, 7 | 1353 | |||
BC126200.1 | - | 6, 7 | 1099 | 8991916 | ||
BC126202.1 | - | 6, 7 | 1099 | 8991918 | ||
BC143902.1 | - | 6, 7 | 1096 | 9052420 |