Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005139.2 | NP_005130.1 | 3 | 377 | ||
GenBank mRNA | AK313945.1 | - | 3 | 152 | ||
BC000871.1 | - | 3 | 209 | 3460409 | ||
BX336217.2 | - | 3 | 377 | |||
CR407648.1 | - | 1 | 67 | |||
M20560.1 | - | 2 | 113 | |||
M63310.1 | - | 2 | 103 |