Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313941.1 | NP_001300870.1 | 6 | 864 | ||
NM_001313943.1 | NP_001300872.1 | 6 | 908 | |||
NM_001313944.1 | NP_001300873.1 | 5 | 709 | |||
NM_001313945.1 | NP_001300874.1 | 5 | 646 | |||
NM_001313946.1 | NP_001300875.1 | 3 | 517 | |||
NM_001313947.1 | NP_001300876.1 | 4 | 648 | |||
NM_001664.3 | NP_001655.1 | 5 | 769 | |||
GenBank mRNA | AF498970.1 | - | 4 | 489 | ||
AK130066.1 | - | 5 | 765 | |||
AK130808.1 | - | 5 | 645 | |||
AK222556.1 | - | 5 | 661 | |||
AK296654.1 | - | 5 | 585 | |||
AK307220.1 | - | 5 | 544 | |||
BC000946.3 | - | 2 | 213 | |||
BC001360.2 | - | 5 | 601 | 3051460 | ||
BC005976.1 | - | 5 | 606 | 4102976 | ||
BT019870.1 | - | 4 | 489 | |||
BU509012.1 | - | 4 | 479 | |||
BX647063.1 | - | 5 | 665 | |||
L09159.1 | - | 4 | 478 | |||
L25080.1 | - | 5 | 640 | |||
LN607834.1 | - | 2 | 215 | |||
X05026.1 | - | 5 | 647 |