Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001698.2 | NP_001689.1 | 4, 5 | 529 | ||
XM_005252066.3 | XP_005252123.1 | 5, 6 | 572 | |||
XM_005252067.4 | XP_005252124.1 | 5, 6 | 574 | |||
XM_005252069.4 | XP_005252126.1 | 5, 6 | 575 | |||
XM_005252072.2 | XP_005252129.1 | 4, 5 | 529 | |||
XM_011518800.2 | XP_011517102.1 | 5, 6 | 576 | |||
XM_011518802.2 | XP_011517104.1 | 5, 6 | 598 | |||
XM_017014849.1 | XP_016870338.1 | 4, 5 | 529 | |||
XM_017014850.1 | XP_016870339.1 | 5, 6 | 669 | |||
XM_017014851.1 | XP_016870340.1 | 5, 6 | 598 | |||
GenBank mRNA | AK302453.1 | - | 4, 5 | 500 | ||
BG706374.1 | - | 4, 5 | 525 | |||
X79888.1 | - | 4, 5 | 498 |