Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164773.1 | NP_001158245.1 | 2 | 99 | ||
NM_001190.3 | NP_001181.2 | 4 | 375 | |||
NM_001284325.1 | NP_001271254.1 | 5 | 856 | |||
GenBank mRNA | AF268047.1 | - | 4 | 324 | ||
AF268048.1 | - | 2 | 48 | |||
AK023909.1 | - | 3 | 1013 | |||
AK093627.1 | - | 5 | 833 | |||
AK223522.1 | - | 4 | 364 | |||
AK309428.1 | - | 4 | 339 | |||
AK310101.1 | - | 2 | 65 | |||
AK310243.1 | - | 5 | 889 | |||
AK314548.1 | - | 4 | 341 | |||
BC001900.1 | - | 4 | 332 | 3537603 | ||
BC004243.1 | - | 3 | 281 | |||
BU178552.1 | - | 379 | ||||
DA129578.1 | - | 4 | 375 | |||
U62739.1 | - | 3 | 295 | |||
U68418.1 | - | 4 | 319 |