Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199.3 | NP_001190.1 | 10, 11 | 1552 | ||
NM_006129.4 | NP_006120.1 | 10, 11 | 1552 | |||
NR_033403.1 | 10, 11 | 1589 | ||||
NR_033404.1 | 10, 11 | 1589 | ||||
XM_006716386.2 | XP_006716449.2 | 10, 11 | 1552 | |||
XM_011544617.1 | XP_011542919.1 | 10, 11 | 1552 | |||
XM_017013738.1 | XP_016869227.1 | 10, 11 | 1552 | |||
GenBank mRNA | AB209590.1 | - | 10, 11 | 1505 | ||
AK291620.1 | - | 10, 11 | 1405 | |||
BC136679.1 | - | 10, 11 | 1368 | 9020669 | ||
BC142953.1 | - | 10, 11 | 1416 | 8860340 | ||
BC144365.1 | - | 10, 11 | 1401 | |||
BC144366.1 | - | 10, 11 | 1405 | |||
M22488.1 | - | 10, 11 | 1315 | |||
U50330.1 | - | 10, 11 | 1286 | |||
Y08723.1 | - | 11, 12 | 1458 | |||
Y08724.1 | - | 10, 11 | 1292 | |||
Y08725.1 | - | 10, 11 | 1292 |