Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242805.2 | NP_001229734.2 | 4, 5 | 757 | ||
NM_001242806.2 | NP_001229735.2 | 3, 4 | 688 | |||
NM_001242810.2 | NP_001229739.2 | 3, 4 | 1044 | |||
NM_001726.4 | NP_001717.3 | 3, 4 | 483 | |||
NM_207189.3 | NP_997072.2 | 3, 4 | 688 | |||
XM_006710853.3 | XP_006710916.1 | 4, 5 | 1273 | |||
XM_006710854.3 | XP_006710917.1 | 5, 6 | 1118 | |||
XM_006710855.3 | XP_006710918.1 | 5, 6 | 1123 | |||
XM_006710856.3 | XP_006710919.1 | 5, 6 | 1097 | |||
XM_006710857.3 | XP_006710920.1 | 5, 6 | 1087 | |||
XM_011542032.2 | XP_011540334.1 | 5, 6 | 607 | |||
XM_011542033.2 | XP_011540335.1 | 5, 6 | 581 | |||
XM_011542034.2 | XP_011540336.1 | 5, 6 | 597 | |||
XM_011542035.2 | XP_011540337.1 | 5, 6 | 571 | |||
XM_011542036.2 | XP_011540338.1 | 3, 4 | 486 | |||
GenBank mRNA | AF019085.1 | - | 3, 4 | 446 | ||
AK303008.1 | - | 3, 4 | 436 | |||
AY338951.1 | - | 4, 5 | 757 | |||
BC005281.1 | - | 3, 4 | 414 | |||
BC017582.1 | - | 3, 4 | 489 | |||
BC047900.1 | - | 3, 4 | 453 | |||
BC062700.1 | - | 3, 4 | 448 | |||
BG717291.1 | - | 3, 4 | 479 | |||
BX648135.1 | - | 3, 4 | 779 | |||
DB072781.1 | - | 3, 4 | 439 | |||
DC401352.1 | - | 3, 4 | 484 | |||
DC402057.1 | - | 3, 4 | 493 |