Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000715.3 | NP_000706.1 | 2 | 281 | ||
XM_005273251.1 | XP_005273308.1 | 2 | 237 | |||
XM_005273252.4 | XP_005273309.1 | 2 | 413 | |||
GenBank mRNA | AK313164.1 | - | 2 | 227 | ||
BC022312.1 | - | 2 | 281 | 4688428 | ||
M31452.1 | - | 2 | 225 | |||
X07853.1 | - | 1 | 95 |