Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014205.3 | NP_055020.1 | 1 | 1071 | ||
GenBank mRNA | AF119497.1 | - | 1 | 1031 | ||
AK027081.1 | - | 1 | 1006 | |||
AK223333.1 | - | 1 | 986 | |||
AK223358.1 | - | 1 | 1041 | |||
BC052240.1 | - | 1 | 1034 | 6107352 | ||
BC104002.2 | - | 1 | 273 | |||
BC104003.2 | - | 1 | 273 | |||
HY200798.1 | - | 1 | 261 |