Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDLRAD4 SIRNA Assays ›
Gene Symbol:
LDLRAD4
Gene Name:
low density lipoprotein receptor class A domain containing 4
Gene Aliases:
C18orf1
Chromosome Location:
Chr.18: 13217691 - 13652754 on Build GRCh38
UniGene ID:
Hs.731853
Species:
Human
siRNA ID
HSS101219
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDLRAD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003674.3
NP_001003674.1
4
1770
NM_001003675.3
NP_001003675.1
3
1716
NM_001276249.1
NP_001263178.1
4
760
NM_001276251.1
NP_001263180.1
3
1304
NM_181481.4
NP_852146.1
6
1106
NM_181482.4
NP_852147.1
6
1311
XM_005258140.1
XP_005258197.1
7
1335
XM_006722353.1
XP_006722416.1
8
1392
XM_006722354.1
XP_006722417.1
6
1196
XM_011525737.1
XP_011524039.1
6
1194
XM_011525738.1
XP_011524040.1
7
1277
XM_017025961.1
XP_016881450.1
7
1315
XM_017025962.1
XP_016881451.1
6
1261
XM_017025963.1
XP_016881452.1
6
1048
XM_017025964.1
XP_016881453.1
6
1049
XM_017025965.1
XP_016881454.1
10
1702
XM_017025966.1
XP_016881455.1
9
1577
XM_017025967.1
XP_016881456.1
9
1574
XM_017025968.1
XP_016881457.1
8
1481
XM_017025969.1
XP_016881458.1
5
2162
XM_017025970.1
XP_016881459.1
5
623
XM_017025971.1
XP_016881460.1
4
1252
XM_017025972.1
XP_016881461.1
3
1225
GenBank mRNA
AF009424.1
-
6
1109
AF009425.1
-
5
1055
AF009426.1
-
3
708
AF009427.1
-
2
654
AK055028.1
-
4
1769
AK291574.1
-
4
632
BC029958.1
-
3
1304
BC030199.1
-
6
1245
5199354
BC066971.1
-
4
679
4794097
BM715555.1
-
1
40
BM991627.1
-
1
416
BX114947.1
-
3
460
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of epithelial to mesenchymal transition
negative regulation of SMAD protein complex assembly
negative regulation of cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of pathway-restricted SMAD protein phosphorylation
R-SMAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0