Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2G SIRNA Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
CAMK, CAMK-II, CAMKG
Chromosome Location:
Chr.10: 73812501 - 73874591 on Build GRCh38
UniGene ID:
Hs.523045
Species:
Human
siRNA ID
HSS101330
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204492.1
NP_001191421.1
3
317
NM_001222.3
NP_001213.2
3
317
NM_001320898.1
NP_001307827.1
3
317
NM_172169.2
NP_751909.1
3
317
NM_172170.4
NP_751910.1
3
317
NM_172171.2
NP_751911.1
3
317
NM_172173.2
NP_751913.1
3
317
XM_005270195.1
XP_005270252.1
3
308
XM_005270197.1
XP_005270254.1
3
308
XM_005270198.1
XP_005270255.1
3
308
XM_005270199.1
XP_005270256.1
3
308
XM_005270200.1
XP_005270257.1
3
308
XM_005270201.1
XP_005270258.1
3
308
XM_005270203.1
XP_005270260.1
3
308
XM_005270205.1
XP_005270262.1
3
308
XM_006717993.1
XP_006718056.1
3
308
XM_006717996.1
XP_006718059.1
3
308
XM_006717997.1
XP_006718060.1
3
308
XM_006717998.1
XP_006718061.1
3
308
XM_011540225.2
XP_011538527.1
3
256
XM_017016716.1
XP_016872205.1
3
308
XM_017016717.1
XP_016872206.1
3
256
XM_017016718.1
XP_016872207.1
3
308
XM_017016719.1
XP_016872208.1
3
308
XM_017016720.1
XP_016872209.1
3
308
XM_017016721.1
XP_016872210.1
3
383
XM_017016722.1
XP_016872211.1
3
308
XM_017016723.1
XP_016872212.1
3
308
XM_017016724.1
XP_016872213.1
3
308
XM_017016725.1
XP_016872214.1
3
308
XM_017016726.1
XP_016872215.1
3
308
XM_017016727.1
XP_016872216.1
3
308
XM_017016728.1
XP_016872217.1
3
256
XM_017016729.1
XP_016872218.1
2
98
XM_017016730.1
XP_016872219.1
3
308
XM_017016731.1
XP_016872220.1
3
269
XM_017016732.1
XP_016872221.1
3
202
XM_017016733.1
XP_016872222.1
3
256
XM_017016734.1
XP_016872223.1
3
295
XM_017016735.1
XP_016872224.1
3
308
XM_017016736.1
XP_016872225.1
3
383
XM_017016737.1
XP_016872226.1
3
265
XM_017016738.1
XP_016872227.1
3
269
XM_017016739.1
XP_016872228.1
3
440
GenBank mRNA
AK095576.1
-
4
414
AK131199.1
-
3
240
AK225771.1
-
3
317
AK291704.1
-
3
309
AK301180.1
-
3
309
BC034044.1
-
3
308
4813948
BU165166.1
-
3
314
DR002605.1
-
3
324
L07044.1
-
3
208
U66064.1
-
3
240
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
MAPK cascade
protein dephosphorylation
calcium ion transport
nervous system development
regulation of skeletal muscle adaptation
insulin secretion
cell differentiation
positive regulation of GTPase activity
protein autophosphorylation
protein oligomerization
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of cellular response to heat
regulation of relaxation of cardiac muscle
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium-dependent protein serine/threonine phosphatase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8c5rs:80/10.239.167.144:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0