Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001226.3 | NP_001217.2 | 6 | 598 | ||
NM_032992.2 | NP_116787.1 | 3 | 331 | |||
NR_133012.1 | 5 | 521 | ||||
GenBank mRNA | BC000305.2 | - | 6 | 546 | 2820468 | |
BC004460.1 | - | 6 | 546 | 2820468 | ||
BM837197.1 | - | 3 | 277 | |||
BX380795.2 | - | 5 | 462 | |||
U20536.1 | - | 6 | 598 | |||
U20537.1 | - | 3 | 253 |