Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001752.3 | NP_001743.1 | 8 | 1047 | ||
GenBank mRNA | AK291585.1 | - | 8 | 1047 | ||
AK301577.1 | - | 7 | 817 | |||
AK310184.1 | - | 7 | 872 | |||
AK315350.1 | - | 8 | 1031 | |||
AY028632.1 | - | 8 | 958 | |||
BC027300.1 | - | 1 | 61 | |||
BC110398.1 | - | 8 | 1036 | 5551309 | ||
BC112217.1 | - | 8 | 996 | 8327685 | ||
BC112219.1 | - | 8 | 996 | 8327687 | ||
X04076.1 | - | 8 | 1028 |