Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000074.2 | NP_000065.1 | 2, 3 | 351 | ||
GenBank mRNA | AF529195.1 | - | 2, 3 | 279 | ||
AK292637.1 | - | 2, 3 | 343 | |||
AY197739.1 | - | 2, 3 | 279 | |||
BC071754.1 | - | 2, 3 | 351 | 30418744 | ||
BC074950.2 | - | 2, 3 | 314 | 30915276 | ||
FN298241.1 | - | 2, 3 | 290 | |||
L07414.1 | - | 2, 3 | 318 | |||
X67878.1 | - | 2, 3 | 324 | |||
X68550.1 | - | 2, 3 | 335 | |||
X96710.1 | - | 2, 3 | 300 | |||
Z15017.1 | - | 2, 3 | 300 |