Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000611.5 | NP_000602.1 | 4 | 459 | ||
NM_001127223.1 | NP_001120695.1 | 3 | 625 | |||
NM_001127225.1 | NP_001120697.1 | 4 | 518 | |||
NM_001127226.1 | NP_001120698.1 | 4 | 453 | |||
NM_001127227.1 | NP_001120699.1 | 4 | 397 | |||
NM_203329.2 | NP_976074.1 | 5 | 575 | |||
NM_203330.2 | NP_976075.1 | 6 | 620 | |||
NM_203331.2 | NP_976076.1 | 5 | 504 | |||
GenBank mRNA | AK128571.1 | - | 2 | 2416 | ||
AK309894.1 | - | 3 | 625 | |||
AK311778.1 | - | 4 | 402 | |||
AL703204.1 | - | 5 | 522 | |||
AU139009.1 | - | 4 | 397 | |||
AU139541.1 | - | 4 | 453 | |||
BC001506.2 | - | 4 | 383 | 2988140 | ||
BI752924.1 | - | 6 | 555 | |||
BQ230563.1 | - | 5 | 423 | |||
BT007104.1 | - | 3 | 343 | |||
CR407661.1 | - | 3 | 343 | |||
CR541722.1 | - | 3 | 343 | |||
DA578905.1 | - | 5 | 519 | |||
DA840870.1 | - | 4 | 519 | |||
DA917093.1 | - | 5 | 486 | |||
M27909.1 | - | 4 | 391 | |||
M34671.1 | - | 3 | 372 | |||
M95708.1 | - | 4 | 405 | |||
X16447.1 | - | 4 | 406 | |||
X17198.1 | - | 4 | 415 |