Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 2, 3 | 734 | ||
NM_001318368.1 | NP_001305297.1 | 2, 3 | 734 | |||
XM_005266216.4 | XP_005266273.1 | 2, 3 | 92 | |||
XM_011534864.1 | XP_011533166.1 | 2, 3 | 713 | |||
GenBank mRNA | AK296875.1 | - | 1, 2 | 77 | ||
AY702976.1 | - | 2, 3 | 196 | |||
BC069634.1 | - | 2, 3 | 219 | 7262421 | ||
BC094784.1 | - | 2, 3 | 734 | |||
BC104492.1 | - | 2, 3 | 222 | 40032854 | ||
BC105950.1 | - | 2, 3 | 222 | 40032852 | ||
BC107601.1 | - | 2, 3 | 221 | 40032849 | ||
X85753.1 | - | 2, 3 | 222 |