- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: CDK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260.2 | NP_001251.1 | 10 | 1505 | ||
NM_001318368.1 | NP_001305297.1 | 10 | 1505 | |||
XM_005266216.4 | XP_005266273.1 | 10 | 863 | |||
XM_011534864.1 | XP_011533166.1 | 9 | 1426 | |||
XM_011534865.2 | XP_011533167.1 | 6 | 1550 | |||
XM_017020317.1 | XP_016875806.1 | 9 | 861 | |||
XM_017020318.1 | XP_016875807.1 | 6 | 595 | |||
GenBank mRNA | AK296084.1 | - | 6 | 495 | ||
AK296875.1 | - | 9 | 848 | |||
AY702976.1 | - | 9 | 909 | |||
BC069634.1 | - | 10 | 990 | 7262421 | ||
BC094784.1 | - | 9 | 1447 | |||
BC104492.1 | - | 10 | 993 | 40032854 | ||
BC105950.1 | - | 10 | 993 | 40032852 | ||
BC107601.1 | - | 10 | 992 | 40032849 | ||
HG518164.1 | - | 4 | 316 | |||
L23208.1 | - | 2 | 103 | |||
X85753.1 | - | 10 | 993 |
Gene Symbol: ADRA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309989.1 | - | 1226 |