Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291484.2 | NP_001278413.1 | 6 | 1542 | ||
NM_001308398.1 | NP_001295327.1 | 6 | 1539 | |||
NM_004363.5 | NP_004354.3 | 6 | 1542 | |||
XM_005258413.3 | XP_005258470.2 | 6 | 1511 | |||
XM_011526322.1 | XP_011524624.1 | 4 | 977 | |||
XM_017026145.1 | XP_016881634.1 | 6 | 1508 | |||
XM_017026146.1 | XP_016881635.1 | 5 | 1256 | |||
GenBank mRNA | AB852566.1 | - | 4 | 861 | ||
AK223101.1 | - | 6 | 1478 | |||
BC034671.1 | - | 6 | 1490 | 5184800 | ||
CR749337.1 | - | 4 | 584 | |||
M15042.1 | - | 6 | 1384 | |||
M16234.1 | - | 2 | 406 | |||
M17191.1 | - | 2 | 452 | |||
M29540.1 | - | 6 | 1509 | |||
X16455.1 | - | 6 | 1381 |