Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320917.1 | NP_001307846.1 | 2 | 967 | ||
NM_012125.3 | NP_036257.1 | 3 | 725 | |||
GenBank mRNA | AB084282.1 | - | 2 | 374 | ||
AF026263.1 | - | 1 | 55 | |||
AF385591.1 | - | 1 | 55 | |||
AF498919.1 | - | 1 | 55 | |||
AK095198.1 | - | 3 | 725 | |||
BC041805.1 | - | 2 | 369 | 5298446 | ||
BC068528.1 | - | 2 | 363 | 30339053 |
Set Membership: |
G-protein coupled receptor |