Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000748.2 | NP_000739.1 | 2 | 386 | ||
XM_017000180.1 | XP_016855669.1 | 2 | 368 | |||
GenBank mRNA | AK313470.1 | - | 2 | 155 | ||
BC075040.2 | - | 2 | 151 | 30915357 | ||
BC075041.2 | - | 2 | 151 | 30915573 | ||
U62437.1 | - | 2 | 386 | |||
X53179.1 | - | 2 | 288 | |||
Y08415.1 | - | 2 | 122 |