Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005502.2 | NP_001005502.1 | 6, 7 | 831 | ||
NM_001874.4 | NP_001865.1 | 6, 7 | 863 | |||
NM_198320.3 | NP_938079.1 | 6, 7 | 849 | |||
GenBank mRNA | AF368463.1 | - | 6, 7 | 821 | ||
AK130146.1 | - | 3, 4 | 439 | |||
AK313180.1 | - | 6, 7 | 821 | |||
BC022276.1 | - | 6, 7 | 821 | 4768771 | ||
DA833423.1 | - | 3, 4 | 390 | |||
J04970.2 | - | 6, 7 | 863 |