Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005212.2 | NP_005203.2 | 4 | 284 | ||
XM_011531620.2 | XP_011529922.1 | 5 | 374 | |||
XM_017007761.1 | XP_016863250.1 | 4 | 306 | |||
GenBank mRNA | M73628.1 | - | 4 | 332 | ||
X66417.1 | - | 4 | 270 |