Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000499.4 | NP_000490.1 | 7 | 1509 | ||
NM_001319216.1 | NP_001306145.1 | 6 | 1425 | |||
NM_001319217.1 | NP_001306146.1 | 7 | 1512 | |||
XM_017021953.1 | XP_016877442.1 | 8 | 1428 | |||
GenBank mRNA | AK223108.1 | - | 7 | 1430 | ||
AK223113.1 | - | 7 | 1427 | |||
AK310810.1 | - | 7 | 1556 | |||
AK313880.1 | - | 7 | 1427 | |||
AM236046.1 | - | 8 | 1313 | |||
AM236047.1 | - | 7 | 608 | |||
AY310359.1 | - | 7 | 1229 | |||
AY871801.1 | - | 6 | 1309 | |||
BC023019.1 | - | 7 | 1394 | 5123393 | ||
K03191.1 | - | 7 | 1399 | |||
M12079.1 | - | 5 | 431 |