Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000102.3 | NP_000093.1 | 3 | 759 | ||
GenBank mRNA | AK094106.1 | - | 3 | 842 | ||
AK289898.1 | - | 3 | 639 | |||
AK308466.1 | - | 3 | 549 | |||
AK312235.1 | - | 3 | 641 | |||
BC062997.1 | - | 3 | 611 | 5171626 | ||
BC063388.1 | - | 3 | 611 | 5170736 | ||
BT020000.1 | - | 3 | 587 | |||
M14564.1 | - | 3 | 628 |