Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DGKB SIRNA Assays ›
Gene Symbol:
DGKB
Gene Name:
diacylglycerol kinase beta
Gene Aliases:
DAGK2, DGK, DGK-BETA
Chromosome Location:
Chr.7: 14145049 - 14974777 on Build GRCh38
UniGene ID:
Hs.567255
Species:
Human
siRNA ID
HSS102628
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DGKB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004080.2
NP_004071.1
18
1726
NM_145695.2
NP_663733.1
18
1726
XM_005249628.2
XP_005249685.1
19
2254
XM_005249629.2
XP_005249686.1
19
2251
XM_005249630.2
XP_005249687.1
18
1767
XM_005249631.3
XP_005249688.1
17
1982
XM_011515153.2
XP_011513455.1
19
2092
XM_011515154.2
XP_011513456.1
19
1808
XM_011515156.2
XP_011513458.1
18
1764
XM_011515157.2
XP_011513459.1
18
1771
XM_011515158.2
XP_011513460.1
17
1907
XM_017011783.1
XP_016867272.1
19
2145
XM_017011784.1
XP_016867273.1
19
1805
XM_017011785.1
XP_016867274.1
18
2039
XM_017011786.1
XP_016867275.1
18
2019
XM_017011787.1
XP_016867276.1
18
1769
XM_017011788.1
XP_016867277.1
18
2055
XM_017011789.1
XP_016867278.1
18
1772
XM_017011790.1
XP_016867279.1
19
2091
XM_017011791.1
XP_016867280.1
19
2090
XM_017011792.1
XP_016867281.1
19
2090
GenBank mRNA
AB018261.1
-
15
1355
AK304719.1
-
17
1982
BC105005.1
-
18
1606
8144008
BC143632.1
-
18
1603
9052145
BC143633.1
-
17
1565
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
platelet activation
intracellular signal transduction
diacylglycerol kinase activity
calcium ion binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0