Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293312.1 | NP_001280241.1 | 12 | 1350 | ||
NM_001349.3 | NP_001340.2 | 13 | 1408 | |||
XM_017003489.1 | XP_016858978.1 | 13 | 1379 | |||
GenBank mRNA | AK129521.1 | - | 13 | 1350 | ||
AK222476.1 | - | 13 | 1238 | |||
AK290607.1 | - | 13 | 1265 | |||
AY762100.1 | - | 13 | 1227 | |||
BC000629.2 | - | 13 | 1241 | 3344322 | ||
BC107749.1 | - | 13 | 1221 | 6709590 | ||
BT006710.1 | - | 13 | 1192 | |||
CR749809.1 | - | 12 | 1158 | |||
J05032.1 | - | 13 | 1282 |