Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001923.4 | NP_001914.3 | 27 | 3652 | ||
GenBank mRNA | AJ002955.1 | - | 27 | 3517 | ||
AK294341.1 | - | 11 | 1426 | |||
AK294493.1 | - | 29 | 3785 | |||
AK299650.1 | - | 26 | 3101 | |||
AK303032.1 | - | 21 | 2534 | |||
AK312436.1 | - | 27 | 3474 | |||
AL831958.1 | - | 16 | 3321 | |||
BC011686.2 | - | 27 | 3481 | 3845478 | ||
BC021044.1 | - | 27 | 3469 | |||
BC032080.1 | - | 13 | 1581 | |||
BC050530.1 | - | 27 | 3545 | 5563018 | ||
BC051764.2 | - | 27 | 3535 | 6063274 | ||
L40326.1 | - | 27 | 3446 | |||
U18299.1 | - | 27 | 3508 | |||
U32986.1 | - | 27 | 3517 |