Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001925.2 | NP_001916.1 | 2, 3 | 289 | ||
GenBank mRNA | BC093959.1 | - | 2, 3 | 213 | 7939804 | |
BC106747.2 | - | 1, 2 | 184 | 40021796 | ||
BC112091.1 | - | 2, 3 | 213 | 8327559 | ||
BU616655.1 | - | 1, 2 | 342 | |||
DN998607.1 | - | 2, 3 | 238 | |||
X65977.1 | - | 2, 3 | 215 |