Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004401.2 | NP_004392.1 | 3 | 428 | ||
NM_213566.1 | NP_998731.1 | 3 | 428 | |||
GenBank mRNA | AF087573.1 | - | 3 | 367 | ||
AF103799.1 | - | 3 | 355 | |||
AK222546.1 | - | 3 | 364 | |||
AK313317.1 | - | 3 | 362 | |||
AU121791.1 | - | 3 | 428 | |||
BC000037.2 | - | 3 | 348 | 3505817 | ||
BC007112.1 | - | 3 | 399 | 4292811 | ||
BC007721.2 | - | 3 | 360 | 4299734 | ||
BT006980.1 | - | 3 | 330 | |||
U91985.1 | - | 3 | 386 |