Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001375.2 | NP_001366.1 | 4 | 536 | ||
GenBank mRNA | AB004574.1 | - | 4 | 462 | ||
AF045937.1 | - | 4 | 466 | |||
AF047016.1 | - | 4 | 462 | |||
AF060222.1 | - | 4 | 483 | |||
AK075449.1 | - | 4 | 451 | |||
AK311145.1 | - | 4 | 460 | |||
AK315358.1 | - | 4 | 478 | |||
BC010419.2 | - | 4 | 414 | 3528108 | ||
BC065209.1 | - | 4 | 481 | 5502061 | ||
BT007047.1 | - | 4 | 390 | |||
BX394955.2 | - | 4 | 587 | |||
FJ752529.1 | - | 4 | 342 |