Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135556.1 | NP_001129028.1 | 7 | 788 | ||
NM_001135557.1 | NP_001129029.1 | 6 | 728 | |||
NM_001278421.1 | NP_001265350.1 | 6 | 779 | |||
NM_001278422.1 | NP_001265351.1 | 6 | 728 | |||
NM_004411.4 | NP_004402.1 | 7 | 839 | |||
XM_011515861.1 | XP_011514163.1 | 7 | 686 | |||
XM_011515862.1 | XP_011514164.1 | 7 | 635 | |||
XM_011515863.2 | XP_011514165.1 | 7 | 793 | |||
XM_017011804.1 | XP_016867293.1 | 7 | 1426 | |||
XM_017011805.1 | XP_016867294.1 | 7 | 613 | |||
XM_017011806.1 | XP_016867295.1 | 7 | 788 | |||
XM_017011807.1 | XP_016867296.1 | 7 | 793 | |||
GenBank mRNA | AF063228.1 | - | 7 | 749 | ||
AF123074.1 | - | 6 | 637 | |||
AK091339.1 | - | 8 | 688 | |||
AK297427.1 | - | 6 | 640 | |||
AK309419.1 | - | 7 | 611 | |||
AK313148.1 | - | 7 | 786 | |||
BC022540.1 | - | 6 | 688 | 4793415 | ||
BC032945.2 | - | 7 | 740 | 4801387 | ||
BC037854.1 | - | 7 | 730 | 4817657 | ||
BC053542.1 | - | 7 | 728 | 5727968 |