Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRDMT1 SIRNA Assays ›
Gene Symbol:
TRDMT1
Gene Name:
tRNA aspartic acid methyltransferase 1
Gene Aliases:
DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
Chromosome Location:
Chr.10: 17136785 - 17202071 on Build GRCh38
UniGene ID:
Hs.351665
Species:
Human
siRNA ID
HSS102864
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRDMT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321006.1
NP_001307935.1
6
1082
NM_001321007.1
NP_001307936.1
9
1455
NM_004412.6
NP_004403.1
8
1225
XM_005252372.4
XP_005252429.1
8
1220
XM_005252374.4
XP_005252431.1
8
1220
XM_005252375.4
XP_005252432.1
6
1082
XM_005252378.3
XP_005252435.1
9
1051
XM_006717388.3
XP_006717451.1
8
1220
XM_011519343.2
XP_011517645.1
7
1154
XM_011519344.2
XP_011517646.1
7
1143
XM_011519345.2
XP_011517647.1
7
1556
XM_017015800.1
XP_016871289.1
6
1082
XM_017015801.1
XP_016871290.1
6
1082
XM_017015802.1
XP_016871291.1
8
3439
XM_017015803.1
XP_016871292.1
9
1053
XM_017015804.1
XP_016871293.1
9
891
XM_017015805.1
XP_016871294.1
8
1088
XM_017015806.1
XP_016871295.1
9
1051
XM_017015807.1
XP_016871296.1
9
1072
XM_017015808.1
XP_016871297.1
9
886
XM_017015809.1
XP_016871298.1
5
1005
XM_017015810.1
XP_016871299.1
8
985
XM_017015811.1
XP_016871300.1
6
951
XM_017015812.1
XP_016871301.1
6
745
GenBank mRNA
AF012128.1
-
8
795
AF045888.1
-
8
794
AJ223333.1
-
8
836
AK291417.1
-
8
794
AK294100.1
-
7
724
AK299027.1
-
6
862
AK303428.1
-
3
442
BC047733.1
-
8
795
6473993
BX537961.1
-
9
1045
CR450316.1
-
8
788
HQ637546.1
-
6
644
HQ637547.1
-
7
711
HQ637548.1
-
6
645
HQ637549.1
-
7
722
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
response to amphetamine
tRNA modification
tRNA methylation
C-5 methylation of cytosine
RNA binding
DNA (cytosine-5-)-methyltransferase activity
tRNA methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-dvnl5:80/10.239.162.83:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0