Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197293.2 | NP_001184222.1 | 8 | 1189 | ||
NM_001244604.1 | NP_001231533.1 | 8 | 831 | |||
NM_001386.5 | NP_001377.1 | 8 | 1172 | |||
GenBank mRNA | AB209195.1 | - | 8 | 910 | ||
AK223557.1 | - | 8 | 1035 | |||
AK291287.1 | - | 8 | 953 | |||
AK299077.1 | - | 8 | 824 | |||
BC056408.1 | - | 8 | 1026 | 6044141 | ||
BC067109.1 | - | 8 | 1056 | 3870039 | ||
BX647115.1 | - | 8 | 1100 | |||
D78013.1 | - | 8 | 1015 | |||
U17279.1 | - | 8 | 812 |